N-Benzyl-2-chloro-N-(2-chloroethyl)ethanamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Benzyl[bis(2-chloroethyl)]amine hydrochloride

043645-500MG 043645-1G 043645-5G
101861-852EA 53.85 USD
101861-852 101861-854 101861-856
N-Benzyl-2-chloro-N-(2-chloroethyl)ethanamine hydrochloride ≥95%
N-Benzyl-2-chloro-N-(2-chloroethyl)ethanamine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆Cl₃N
MW: 268.62 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD00185654
CAS Number: 10429-82-0

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR