1,3-Benzoxazol-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-1,3-benzoxazole

043643-500MG 043643-1G 043643-5G
101861-840EA 107.69 USD
101861-840 101861-842 101861-844
1,3-Benzoxazol-4-amine ≥95%
1,3-Benzoxazol-4-amine

Hazard: Irritant

Formula: C₇H₆N₂O
Melting Pt: 77…79 °C
CAS Number: 163808-09-1

Order Now

Specification Test Results

Melting point 77-79°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR