1,3-Benzoxazol-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Aminobenzoxazole, 5-Amino-1,3-benzoxazole

043642-1G 043642-5G
101861-836EA 373.85 USD
101861-836 101861-838
1,3-Benzoxazol-5-amine ≥95%
1,3-Benzoxazol-5-amine

Hazard: Irritant/Store in Freezer

Formula: C₇H₆N₂O
MW: 134.135 g/mol
Melting Pt: 64…66 °C
MDL Number: MFCD05256201
CAS Number: 63837-12-7

Order Now

Specification Test Results

Melting point 64-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR