Ethyl-3-{[3-(trifluoromethyl)benzoyl]amino}-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-[3-(trifluoromethyl)benzamido]-1H-indole-2-carboxylate

043637-500MG 043637-1G 043637-5G
101861-814EA 747.69 USD
101861-814 101861-816 101861-818
Ethyl-3-{[3-(trifluoromethyl)benzoyl]amino}-1H-indole-2-carboxylate ≥95%
Ethyl-3-{[3-(trifluoromethyl)benzoyl]amino}-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₉H₁₅F₃N₂O₃
MW: 376.34 g/mol
Melting Pt: 212…214 °C
MDL Number: MFCD00794680
CAS Number: 338968-28-8

Order Now

Specification Test Results

Melting point 212-214°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR