5-[2-(Methylsulfanyl)phenyl]-1,3,4-oxadiazol-2-ylhydrosulfide ≥95%

Supplier: Matrix Scientific

043636-500MG 043636-1G 043636-5G
101861-808EA 375.38 USD
101861-808 101861-810 101861-812
5-[2-(Methylsulfanyl)phenyl]-1,3,4-oxadiazol-2-ylhydrosulfide ≥95%
5-[2-(Methylsulfanyl)phenyl]-1,3,4-oxadiazol-2-ylhydrosulfide

Hazard: Irritant

Formula: C₉H₈N₂OS₂
Melting Pt: 230…231 °C
MDL Number: MFCD00974931
CAS Number: 338968-03-9

Order Now

Specification Test Results

Melting point 230-231°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR