6-[(2-Chlorophenyl)sulfanyl]nicotinaldehyde ≥95%

Supplier: Matrix Scientific

043635-500MG 043635-1G 043635-5G
101861-802EA 747.69 USD
101861-802 101861-804 101861-806
6-[(2-Chlorophenyl)sulfanyl]nicotinaldehyde ≥95%
6-[(2-Chlorophenyl)sulfanyl]nicotinaldehyde

Hazard: Irritant

Formula: C₁₂H₈ClNOS
MW: 249.72 g/mol
Melting Pt: 65…66.5 °C
MDL Number: MFCD00974921
CAS Number: 338967-22-9

Order Now

Specification Test Results

Melting point 65-66.5°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR