6-[3-(Trifluoromethyl)phenoxy]nicotinaldehyde ≥95%

Supplier: Matrix Scientific

043634-500MG 043634-1G 043634-5G
101861-796EA 747.69 USD
101861-796 101861-798 101861-800
6-[3-(Trifluoromethyl)phenoxy]nicotinaldehyde ≥95%
6-[3-(Trifluoromethyl)phenoxy]nicotinaldehyde

Hazard: Irritant

Formula: C₁₃H₈F₃NO₂
MW: 267.21 g/mol
Melting Pt: 51…54 °C
MDL Number: MFCD00974920
CAS Number: 338967-16-1

Order Now

Specification Test Results

Melting point 51-54°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR