2-Amino-2-(4-bromophenyl)acetic acid ≥95%
Supplier: Matrix Scientific
043633-500MG
043633-1G
043633-5G
101861-790EA
138.46
USD
101861-790
101861-792
101861-794
2-Amino-2-(4-bromophenyl)acetic acid ≥95%
2-Amino-2-(4-bromophenyl)acetic acid
Hazard: Irritant
Formula:
C₈H₈BrNO₂ Melting Pt: 273…274 °C |
MDL Number:
MFCD00049327 CAS Number: 71079-03-3 |
Specification Test Results
Melting point | 273-274°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...