2-Amino-2-(3-bromophenyl)acetic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Bromo-DL-phenylglycine
043632-500MG
043632-1G
101861-786EA
335.38
USD
101861-786
101861-788
2-Amino-2-(3-bromophenyl)acetic acid ≥95%
2-Amino-2-(3-bromophenyl)acetic acid
Hazard: Irritant
Formula:
C₈H₈BrNO₂ MW: 230.0600 g/mol Melting Pt: 212-215°C |
MDL Number:
MFCD00665358 CAS Number: 79422-73-4 |
Specification Test Results
Melting point | 212-215°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...