6-Nitro-2,3-dihydro-1H-inden-5-ylamine ≥95%
Supplier: Matrix Scientific
043628-500MG
043628-1G
043628-5G
101861-768EA
556.92
USD
101861-768
101861-770
101861-772
6-Nitro-2,3-dihydro-1H-inden-5-ylamine ≥95%
6-Nitro-2,3-dihydro-1H-inden-5-ylamine
Hazard: Irritant
Formula:
C₉H₁₀N₂O₂ Melting Pt: 126…128 °C |
MDL Number:
MFCD00172125 CAS Number: 52957-66-1 |
Specification Test Results
Melting point | 126-128°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...