4-Methoxy-N-[1-(methylsulfanyl)-2-nitrovinyl]aniline ≥95%

Supplier: Matrix Scientific

043626-500MG 043626-1G
101861-758EA 278.46 USD
101861-758 101861-760
4-Methoxy-N-[1-(methylsulfanyl)-2-nitrovinyl]aniline ≥95%
4-Methoxy-N-[1-(methylsulfanyl)-2-nitrovinyl]aniline

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃S
Melting Pt: 157…158.5 °C
MDL Number: MFCD00138259
CAS Number: 62390-80-1

Order Now

Specification Test Results

Melting point 157-158.5°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR