Ethyl-4-acetyl-5-anilino-3-methyl-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

043619-500MG 043619-1G 043619-5G
101861-730EA 181.54 USD
101861-730 101861-732 101861-734
Ethyl-4-acetyl-5-anilino-3-methyl-2-thiophenecarboxylate ≥95%
Ethyl-4-acetyl-5-anilino-3-methyl-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₆H₁₇NO₃S
Melting Pt: 123…124 °C
MDL Number: MFCD03414171
CAS Number: 393802-93-2

Order Now

Specification Test Results

Melting point 123-124°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR