{4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanol ≥95%

Supplier: Matrix Scientific

043618-500MG 043618-1G 043618-5G
101861-724EA 158.46 USD
101861-724 101861-726 101861-728
{4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanol ≥95%
{4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanol

Hazard: Irritant

Formula: C₁₂H₁₀F₃NOS
Melting Pt: 120…123 °C
MDL Number: MFCD03791170
CAS Number: 317318-96-0

Order Now

Specification Test Results

Melting point 120-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR