Dimethyl-2-[2-nitro-4-(trifluoromethyl)phenyl]malonate ≥95%

Supplier: Matrix Scientific

Synonyms: Dimethyl 2-(2-nitro-4-(trifluoromethyl)phenyl)malonate

043616-500MG 043616-1G 043616-5G
101861-718EA 64.62 USD
101861-718 101861-720 101861-722
Dimethyl-2-[2-nitro-4-(trifluoromethyl)phenyl]malonate ≥95%
Dimethyl-2-[2-nitro-4-(trifluoromethyl)phenyl]malonate

Hazard: Irritant

Formula: C₁₂H₁₀F₃NO₆
MW: 321.21 g/mol
Melting Pt: 57…58 °C
MDL Number: MFCD03617804
CAS Number: 290825-52-4

Order Now

Specification Test Results

Melting point 57-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR