2-Chloro-4-morpholinobenzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-morpholinobenzoic acid

043613-500MG 043613-1G 043613-5G
101861-704EA 246.15 USD
101861-704 101861-706 101861-708
2-Chloro-4-morpholinobenzenecarboxylic acid ≥95%
2-Chloro-4-morpholinobenzenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₃
MW: 241.68 g/mol
Melting Pt: 188.5…192 °C
MDL Number: MFCD03425732
CAS Number: 175153-55-6

Order Now

Specification Test Results

Melting point 188.5-192°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR