2-Chloro-4-(1-pyrrolidinyl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

043610-500MG 043610-1G 043610-5G
101861-686EA 375.38 USD
101861-686 101861-688 101861-690
2-Chloro-4-(1-pyrrolidinyl)benzenecarboxylic acid ≥95%
2-Chloro-4-(1-pyrrolidinyl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₂
Melting Pt: 232…234 °C
MDL Number: MFCD03001210
CAS Number: 192513-60-3

Order Now

Specification Test Results

Melting point 232-234°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR