Methyl-2-chloro-4-(1-pyrrolidinyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-Chloro-4-(1-pyrrolidinyl)benzoate, Methyl 2-chloro-4-(pyrrolidin-1-yl)benzoate

043609-500MG 043609-1G 043609-5G
101861-680EA 375.38 USD
101861-680 101861-682 101861-684
Methyl-2-chloro-4-(1-pyrrolidinyl)benzenecarboxylate ≥95%
Methyl-2-chloro-4-(1-pyrrolidinyl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₄ClNO₂
MW: 239.7 g/mol
Melting Pt: 133…135 °C
MDL Number: MFCD03844850
CAS Number: 175153-38-5

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR