2-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carbohydrazide ≥95%

Supplier: Matrix Scientific

043606-500MG 043606-1G 043606-5G
101861-668EA 375.38 USD
101861-668 101861-670 101861-672
2-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carbohydrazide ≥95%
2-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carbohydrazide

Hazard: Irritant

Formula: C₉H₈F₃N₅OS
Melting Pt: 206…208 °C
MDL Number: MFCD03305519
CAS Number: 955964-92-8

Order Now

Specification Test Results

Melting point 206-208°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR