2-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

043605-500MG 043605-1G 043605-5G
101861-662EA 278.46 USD
101861-662 101861-664 101861-666
2-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid ≥95%
2-[3-Methyl-5-(trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₉H₆F₃N₃O₂S
Melting Pt: 170…172 °C
MDL Number: MFCD03012088
CAS Number: 449778-74-9

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR