2-(4,4,4-Trifluoro-1-methyl-3-oxobutylidene)-1-hydrazinecarbothioamide ≥95%

Supplier: Matrix Scientific

043604-500MG 043604-1G 043604-5G
101861-656EA 181.54 USD
101861-656 101861-658 101861-660
2-(4,4,4-Trifluoro-1-methyl-3-oxobutylidene)-1-hydrazinecarbothioamide ≥95%
2-(4,4,4-Trifluoro-1-methyl-3-oxobutylidene)-1-hydrazinecarbothioamide

Hazard: Irritant

Formula: C₆H₈F₃N₃OS
Melting Pt: 144…145 °C
MDL Number: MFCD03011851
CAS Number: 383148-57-0

Order Now

Specification Test Results

Melting point 144-145°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR