4-(3-Chloro-2-thienyl)-2-pyrimidinamine ≥95%

Supplier: Matrix Scientific

043602-500MG 043602-1G
101861-648EA 556.92 USD
101861-648 101861-650
4-(3-Chloro-2-thienyl)-2-pyrimidinamine ≥95%
4-(3-Chloro-2-thienyl)-2-pyrimidinamine

Hazard: Irritant

Formula: C₈H₆ClN₃S
Melting Pt: 152…153 °C
MDL Number: MFCD03791168
CAS Number: 855308-67-7

Order Now

Specification Test Results

Melting point 152-153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR