3-Amino-3-[4-(methoxycarbonyl)phenyl]propanoic acid ≥95%
Supplier: Matrix Scientific
043601-500MG
043601-1G
043601-5G
101861-642EA
278.46
USD
101861-642
101861-644
101861-646
3-Amino-3-[4-(methoxycarbonyl)phenyl]propanoic acid ≥95%
3-Amino-3-[4-(methoxycarbonyl)phenyl]propanoic acid
Hazard: Irritant
Formula:
C₁₁H₁₃NO₄ MW: 223.23 g/mol Melting Pt: 230…232 °C |
MDL Number:
MFCD03002438 CAS Number: 181519-49-3 |
Specification Test Results
Melting point | 230-232°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...