4-(5-Bromo-2-thienyl)-2-pyrimidinamine ≥95%

Supplier: Matrix Scientific

043600-500MG 043600-1G 043600-5G
101861-636EA 375.38 USD
101861-636 101861-638 101861-640
4-(5-Bromo-2-thienyl)-2-pyrimidinamine ≥95%
4-(5-Bromo-2-thienyl)-2-pyrimidinamine

Hazard: Irritant

Formula: C₈H₆BrN₃S
Melting Pt: 189…191 °C
MDL Number: MFCD03617980
CAS Number: 855308-66-6

Order Now

Specification Test Results

Melting point 189-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR