4-(5-Chloro-2-thienyl)-2-pyrimidinamine ≥95%

Supplier: Matrix Scientific

043599-500MG 043599-1G 043599-5G
101861-630EA 375.38 USD
101861-630 101861-632 101861-634
4-(5-Chloro-2-thienyl)-2-pyrimidinamine ≥95%
4-(5-Chloro-2-thienyl)-2-pyrimidinamine

Hazard: Irritant

Formula: C₈H₆ClN₃S
Melting Pt: 184…186 °C
MDL Number: MFCD03617979
CAS Number: 855308-65-5

Order Now

Specification Test Results

Melting point 184-186°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR