1-(5-Chloro-2-thienyl)-3-(dimethylamino)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

043598-500MG 043598-1G 043598-5G
101861-624EA 181.54 USD
101861-624 101861-626 101861-628
1-(5-Chloro-2-thienyl)-3-(dimethylamino)-2-propen-1-one ≥95%
1-(5-Chloro-2-thienyl)-3-(dimethylamino)-2-propen-1-one

Hazard: Irritant

Formula: C₉H₁₀ClNOS
Melting Pt: 118…119 °C
MDL Number: MFCD03617978
CAS Number: 166196-78-7

Order Now

Specification Test Results

Melting point 118-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR