1-(5-Bromo-2-thienyl)-3-(dimethylamino)-2-propen-1-one ≥95%
Supplier: Matrix Scientific
043597-500MG
043597-1G
043597-5G
101861-618EA
181.54
USD
101861-618
101861-620
101861-622
1-(5-Bromo-2-thienyl)-3-(dimethylamino)-2-propen-1-one ≥95%
1-(5-Bromo-2-thienyl)-3-(dimethylamino)-2-propen-1-one
Hazard: Irritant
Formula:
C₉H₁₀BrNOS Melting Pt: 117…118 °C |
MDL Number:
MFCD00098108 CAS Number: 938176-46-6 |
Specification Test Results
Melting point | 117-118°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...