4-Methyl-6-(2-thienyl)-2-pyrimidinamine ≥95%

Supplier: Matrix Scientific

043596-500MG 043596-1G 043596-5G
101861-612EA 375.38 USD
101861-612 101861-614 101861-616
4-Methyl-6-(2-thienyl)-2-pyrimidinamine ≥95%
4-Methyl-6-(2-thienyl)-2-pyrimidinamine

Hazard: Irritant

Formula: C₉H₉N₃S
MW: 191.26 g/mol
Melting Pt: 171…172 °C
MDL Number: MFCD03787266
CAS Number: 26963-43-9

Order Now

Specification Test Results

Melting point 171-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR