3-Amino-3-(4-cyanophenyl)propanoic acid ≥95%
Supplier: Matrix Scientific
043595-500MG
043595-1G
043595-5G
101861-606EA
150.77
USD
101861-606
101861-608
101861-610
3-Amino-3-(4-cyanophenyl)propanoic acid ≥95%
3-Amino-3-(4-cyanophenyl)propanoic acid
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₂ Melting Pt: 226…228 °C (dec.) |
MDL Number:
MFCD01871335 CAS Number: 80971-95-5 |
Specification Test Results
Melting point | 226-228°C(d |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...