4-(4-Methylpiperazino)-1,2-benzenediamine ≥95%
Supplier: Matrix Scientific
043593-500MG
043593-1G
043593-5G
101861-594EA
492.31
USD
101861-594
101861-596
101861-598
4-(4-Methylpiperazino)-1,2-benzenediamine ≥95%
4-(4-Methylpiperazino)-1,2-benzenediamine
Hazard: Irritant
Formula:
C₁₁H₁₈N₄ Melting Pt: 118…120 °C |
MDL Number:
MFCD03001209 CAS Number: 54998-08-2 |
Specification Test Results
Melting point | 118-120°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...