1-Methyl-5-(4-methylphenoxy)-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

043587-500MG 043587-1G 043587-5G
101861-560EA 747.69 USD
101861-560 101861-562 101861-564
1-Methyl-5-(4-methylphenoxy)-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde ≥95%
1-Methyl-5-(4-methylphenoxy)-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₁F₃N₂O₂
Melting Pt: 83…85 °C
MDL Number: MFCD03425731
CAS Number: 449778-73-8

Order Now

Specification Test Results

Melting point 83-85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR