5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(4-Hydroxypiperidin-1-yl)thiophene-2-carbaldehyde, 5-(4-Hydroxypiperidin-1-yl)thiophene-2-carboxaldehyde, 5-(4-Hydroxypiperidino)thiophene-2-carboxaldehyde

043583-500MG 043583-1G 043583-5G
101861-538EA 60 USD
101861-538 101861-540 101861-542
5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde ≥95%
5-(4-Hydroxypiperidino)-2-thiophenecarbaldehyde

Hazard: Irritant

Formula: C₁₀H₁₃NO₂S
MW: 211.28 g/mol
Melting Pt: 137…139 °C
MDL Number: MFCD03425728
CAS Number: 207290-72-0

Order Now

Specification Test Results

Melting point 137-139°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR