5-Amino-1-phenyl-1H-pyrazole-4-carbohydrazide ≥95%
Supplier: Matrix Scientific
043582-500MG
043582-1G
043582-5G
101861-532EA
181.54
USD
101861-532
101861-534
101861-536
5-Amino-1-phenyl-1H-pyrazole-4-carbohydrazide ≥95%
5-Amino-1-phenyl-1H-pyrazole-4-carbohydrazide
Hazard: Irritant
Formula:
C₁₀H₁₁N₅O Melting Pt: 188…190 °C |
MDL Number:
MFCD03056132 CAS Number: 58046-54-1 |
Specification Test Results
Melting point | 188-190°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...