2-Acetylbenzonitrile 95

Supplier: Matrix Scientific

Synonyms: 2-Acetylbenzenecarbonitrile

043581-1G 043581-5G
101861-530EA 75.38 USD
101861-530 102551-326
2-Acetylbenzonitrile 95
2-Acetylbenzonitrile

Hazard: Irritant

Formula: C₉H₇NO
MW: 145.1600 g/mol
Melting Pt: 46…48 °C
MDL Number: MFCD00272571
CAS Number: 91054-33-0

Order Now

Specification Test Results

Melting point 46-48°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR