6-(2-Fluorophenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

043577-500MG 043577-1G 043577-5G
101861-514EA 375.38 USD
101861-514 101861-516 101861-518
6-(2-Fluorophenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde ≥95%
6-(2-Fluorophenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₇FN₂OS
MW: 246.26 g/mol
Melting Pt: 147…148 °C
MDL Number: MFCD02091516
CAS Number: 439095-02-0

Order Now

Specification Test Results

Melting point 147-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR