2-Chloro-6-methylquinoline-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-methylquinoline-3-carboxaldehyde, 2-Chloro-3-formyl-6-methylquinoline

043576-500MG 043576-1G
101861-510EA 375.38 USD
101861-510 101861-512
2-Chloro-6-methylquinoline-3-carbaldehyde ≥95%
2-Chloro-6-methylquinoline-3-carbaldehyde

Hazard: Irritant

Formula: C₁₁H₈ClNO
MW: 205.6400 g/mol
Melting Pt: 123 °C
MDL Number: MFCD01912773
CAS Number: 73568-27-1

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR