tert-Butyl-N-(1-benzyl-2-oxoethyl)carbamate ≥95%
Supplier: Matrix Scientific
043575-500MG
043575-1G
101861-506EA
747.69
USD
101861-506
101861-508
tert-Butyl-N-(1-benzyl-2-oxoethyl)carbamate ≥95%
tert-Butyl-N-(1-benzyl-2-oxoethyl)carbamate
Hazard: Irritant
Formula:
C₁₄H₁₉NO₃ MW: 249.31 g/mol Melting Pt: 68…70 °C |
MDL Number:
MFCD03425726 CAS Number: 103127-53-3 |
Specification Test Results
Melting point | 68-70°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...