8-Benzyl-2,8-diazaspiro[4.5]decane-1,3-dione ≥95%
Supplier: Matrix Scientific
043568-500MG
043568-1G
101861-484EA
700
USD
101861-484
101861-486
8-Benzyl-2,8-diazaspiro[4.5]decane-1,3-dione ≥95%
8-Benzyl-2,8-diazaspiro[4.5]decane-1,3-dione
Hazard: Irritant
Formula:
C₁₅H₁₈N₂O₂ Melting Pt: 189 °C |
MDL Number:
MFCD11043250 CAS Number: 1463-48-5 |
Specification Test Results
Melting point | 189°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...