Ethyl-2-(1-benzyl-4-piperidinylidene)-2-cyanoacetate ≥95%
Supplier: Matrix Scientific
043567-500MG
043567-1G
043567-5G
101861-478EA
278.46
USD
101861-478
101861-480
101861-482
Ethyl-2-(1-benzyl-4-piperidinylidene)-2-cyanoacetate ≥95%
Ethyl-2-(1-benzyl-4-piperidinylidene)-2-cyanoacetate
Hazard: Irritant
Formula:
C₁₇H₂₀N₂O₂ Melting Pt: 64…67 °C |
MDL Number:
MFCD01569592 CAS Number: 1463-52-1 |
Specification Test Results
Melting point | 64-67°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...