1-(3,4-Dichlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
Supplier: Matrix Scientific
043560-500MG
043560-1G
043560-5G
101861-448EA
278.46
USD
101861-448
101861-450
101861-452
1-(3,4-Dichlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid ≥95%
1-(3,4-Dichlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₀Cl₂N₂O₂ Melting Pt: 238…240 °C |
MDL Number:
MFCD02102423 CAS Number: 477710-54-6 |
Specification Test Results
Melting point | 238-240°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...