Ethyl-1-(5-amino-2-pyridinyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
043556-500MG
043556-1G
043556-5G
101861-430EA
375.38
USD
101861-430
101861-432
101861-434
Ethyl-1-(5-amino-2-pyridinyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%
Ethyl-1-(5-amino-2-pyridinyl)-3,5-dimethyl-1H-pyrazole-4-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₆N₄O₂ Melting Pt: 137…140 °C |
MDL Number:
MFCD02571875 CAS Number: 477710-44-4 |
Specification Test Results
Melting point | 137-140°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...