Methyl-3-nitro-4-(phenylsulfonyl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

043555-500MG 043555-1G 043555-5G
101861-424EA 278.46 USD
101861-424 101861-426 101861-428
Methyl-3-nitro-4-(phenylsulfonyl)benzenecarboxylate ≥95%
Methyl-3-nitro-4-(phenylsulfonyl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₁₁NO₆S
Melting Pt: 137…138 °C
MDL Number: MFCD00706470
CAS Number: 306278-46-6

Order Now

Specification Test Results

Melting point 137-138°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR