2-Methyl-2H-indazol-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-methylindazole, 5-Amino-2-methyl-2H-indazole

043554-500MG 043554-1G
101861-420EA 1109.23 USD
101861-420 101861-422
2-Methyl-2H-indazol-5-amine ≥95%
2-Methyl-2H-indazol-5-amine

Hazard: Irritant

Formula: C₈H₉N₃
MW: 147.18 g/mol
Melting Pt: 125 °C
MDL Number: MFCD03787060
CAS Number: 60518-59-4

Order Now

Specification Test Results

Melting point 125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR