2,3-Dimethyl-6-nitroquinoxaline ≥95%

Supplier: Matrix Scientific

043553-500MG 043553-1G 043553-5G
101861-414EA 181.54 USD
101861-414 101861-416 101861-418
2,3-Dimethyl-6-nitroquinoxaline ≥95%
2,3-Dimethyl-6-nitroquinoxaline

Hazard: Irritant

Formula: C₁₀H₉N₃O₂
Melting Pt: 130-131°C
MDL Number: MFCD00205436
CAS Number: 2942-03-2

Order Now

Specification Test Results

Melting point 130-131°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR