6-Amino-5-bromonicotinonitrile ≥95%

Supplier: Matrix Scientific

043552-500MG 043552-1G 043552-5G
101861-408EA 112.31 USD
101861-408 101861-410 101861-412
6-Amino-5-bromonicotinonitrile ≥95%
6-Amino-5-bromonicotinonitrile

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
Melting Pt: 194…195 °C
MDL Number: MFCD02102412
CAS Number: 477871-32-2

Order Now

Specification Test Results

Melting point 194-195°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR