3-(1H-Pyrrol-1-yl)-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

043551-500MG 043551-1G 043551-5G
101861-402EA 375.38 USD
101861-402 101861-404 101861-406
3-(1H-Pyrrol-1-yl)-2-thiophenecarboxylic acid ≥95%
3-(1H-Pyrrol-1-yl)-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₉H₇NO₂S
Melting Pt: 184 °C (dec.)
MDL Number: MFCD00100445
CAS Number: 74772-17-1

Order Now

Specification Test Results

Melting point 184°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR