1,2-Dihydronaphtho[2,1-b]furan-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

043549-500MG 043549-1G 043549-5G
101861-390EA 300 USD
101861-390 101861-392 101861-394
1,2-Dihydronaphtho[2,1-b]furan-2-carboxylic acid ≥95%
1,2-Dihydronaphtho[2,1-b]furan-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₀O₃
Melting Pt: 330 °C
MDL Number: MFCD03001250
CAS Number: 24758-31-4

Order Now

Specification Test Results

Melting point 330°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR