Ethyl-1,2-dihydronaphtho[2,1-b]furan-2-carboxylate ≥95%

Supplier: Matrix Scientific

043548-500MG 043548-1G 043548-5G
101861-384EA 278.46 USD
101861-384 101861-386 101861-388
Ethyl-1,2-dihydronaphtho[2,1-b]furan-2-carboxylate ≥95%
Ethyl-1,2-dihydronaphtho[2,1-b]furan-2-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MW: 242.28 g/mol
Melting Pt: 64…66 °C
MDL Number: MFCD03001249
CAS Number: 62019-34-5

Order Now

Specification Test Results

Melting point 64-66°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR