1-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1-ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4'-(2-Chloro-6-fluorobenzyloxy)acetophenone

043546-500MG 043546-1G 043546-5G
101861-372EA 89.23 USD
101861-372 101861-374 101861-376
1-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1-ethanone ≥95%
1-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1-ethanone

Hazard: Irritant

Formula: C₁₅H₁₂ClFO₂
MW: 278.71 g/mol
Melting Pt: 91…93 °C
MDL Number: MFCD02179827
CAS Number: 306934-77-0

Order Now

Specification Test Results

Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR