3-Oxo-3-[3-(1H-pyrrol-1-yl)-2-thienyl]propanenitrile ≥95%

Supplier: Matrix Scientific

043545-500MG 043545-1G 043545-5G
101861-366EA 375.38 USD
101861-366 101861-368 101861-370
3-Oxo-3-[3-(1H-pyrrol-1-yl)-2-thienyl]propanenitrile ≥95%
3-Oxo-3-[3-(1H-pyrrol-1-yl)-2-thienyl]propanenitrile

Hazard: Irritant

Formula: C₁₁H₈N₂OS
MW: 216.26 g/mol
Melting Pt: 99…101 °C
MDL Number: MFCD02102396
CAS Number: 477871-05-9

Order Now

Specification Test Results

Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR