Methyl-3-(1H-pyrrol-1-yl)-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

043543-500MG 043543-1G 043543-5G
101861-360EA 278.46 USD
101861-360 101861-362 101861-364
Methyl-3-(1H-pyrrol-1-yl)-2-thiophenecarboxylate ≥95%
Methyl-3-(1H-pyrrol-1-yl)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₂S
Melting Pt: 63…64 °C
MDL Number: MFCD00053052
CAS Number: 74772-16-0

Order Now

Specification Test Results

Melting point 63-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR